Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aurkc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080965.1 | NP_001074434.1 | 4 - 5 | 529 | 103 | |
NM_001080966.1 | NP_001074435.1 | 4 - 5 | 815 | 103 | ||
NM_020572.2 | NP_065597.2 | 4 - 5 | 693 | 103 | ||
GenBank mRNA | BC064780.1 | - | 4 - 5 | 697 | 30476803 | 103 |
BC100337.1 | - | 4 - 5 | 450 | 103 | ||
BG080448.2 | - | 4 - 5 | 506 | 103 | ||
DV648982.1 | - | 4 - 5 | 557 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |