Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033294.3 | NP_001028466.2 | 11 - 12 | 1353 | 80 | |
GenBank mRNA | AK144739.1 | - | 11 - 12 | 1066 | 80 | |
AK166327.1 | - | 11 - 12 | 1023 | 80 | ||
BC066017.1 | - | 11 - 12 | 990 | 6401415 | 80 | |
BC132351.1 | - | 11 - 12 | 1043 | 40130628 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |