Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033294.3 | NP_001028466.2 | 18 - 19 | 2177 | 69 | |
GenBank mRNA | AK030888.1 | - | 18 - 19 | 2124 | 69 | |
AK144739.1 | - | 18 - 19 | 1891 | 69 | ||
AK166327.1 | - | 18 - 19 | 1847 | 69 | ||
BC066017.1 | - | 18 - 19 | 1814 | 6401415 | 69 | |
BC132351.1 | - | 18 - 19 | 1867 | 40130628 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |