Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200003535.mm.1 |
NM_001033294.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200003535.mm.1 |
NM_001033294.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200003535.mm.1 |
TC0200000495.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200003539.mm.1 |
NM_001033294.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200003539.mm.1 |
NM_001033294.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200003539.mm.1 |
TC0200000495.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200003536.mm.1 |
NM_001033294.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200003536.mm.1 |
NM_001033294.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200003536.mm.1 |
TC0200000495.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200003538.mm.1 |
NM_001033294.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200003538.mm.1 |
NM_001033294.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200003538.mm.1 |
TC0200000495.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200003540.mm.1 |
NM_001033294.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200003540.mm.1 |
NM_001033294.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200003540.mm.1 |
TC0200000495.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200003537.mm.1 |
NM_001033294.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200003537.mm.1 |
NM_001033294.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200003537.mm.1 |
TC0200000495.mm.1 |
MTA TC |