Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200008935.mm.1 |
NM_029056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008935.mm.1 |
NM_029056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008935.mm.1 |
TC1200001252.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008938.mm.1 |
NM_029056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008938.mm.1 |
NM_029056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008938.mm.1 |
TC1200001252.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008941.mm.1 |
NM_029056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008941.mm.1 |
NM_029056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008941.mm.1 |
TC1200001252.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008956.mm.1 |
NM_029056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008956.mm.1 |
NM_029056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008956.mm.1 |
TC1200001252.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008946.mm.1 |
NM_029056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008946.mm.1 |
NM_029056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008946.mm.1 |
TC1200001252.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008937.mm.1 |
NM_029056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008937.mm.1 |
NM_029056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008937.mm.1 |
TC1200001252.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008945.mm.1 |
NM_029056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008945.mm.1 |
NM_029056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008945.mm.1 |
TC1200001252.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008953.mm.1 |
NM_029056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008953.mm.1 |
NM_029056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008953.mm.1 |
TC1200001252.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008954.mm.1 |
NM_029056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008954.mm.1 |
NM_029056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008954.mm.1 |
TC1200001252.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008955.mm.1 |
NM_029056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008955.mm.1 |
NM_029056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008955.mm.1 |
TC1200001252.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008933.mm.1 |
NM_029056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008933.mm.1 |
NM_029056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008933.mm.1 |
TC1200001252.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008942.mm.1 |
NM_029056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008942.mm.1 |
NM_029056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008942.mm.1 |
TC1200001252.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008950.mm.1 |
NM_029056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008950.mm.1 |
NM_029056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008950.mm.1 |
TC1200001252.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008952.mm.1 |
NM_029056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008952.mm.1 |
NM_029056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008952.mm.1 |
TC1200001252.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008940.mm.1 |
NM_029056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008940.mm.1 |
NM_029056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008940.mm.1 |
TC1200001252.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008947.mm.1 |
NM_029056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008947.mm.1 |
NM_029056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008947.mm.1 |
TC1200001252.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008949.mm.1 |
NM_029056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008949.mm.1 |
NM_029056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008949.mm.1 |
TC1200001252.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008958.mm.1 |
NM_029056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008958.mm.1 |
NM_029056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008958.mm.1 |
TC1200001252.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008948.mm.1 |
NM_029056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008948.mm.1 |
NM_029056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008948.mm.1 |
TC1200001252.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008951.mm.1 |
NM_029056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008951.mm.1 |
NM_029056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008951.mm.1 |
TC1200001252.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008959.mm.1 |
NM_029056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008959.mm.1 |
NM_029056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008959.mm.1 |
TC1200001252.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008944.mm.1 |
NM_029056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008944.mm.1 |
NM_029056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008944.mm.1 |
TC1200001252.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008934.mm.1 |
NM_029056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008934.mm.1 |
NM_029056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008934.mm.1 |
TC1200001252.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008939.mm.1 |
NM_029056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008939.mm.1 |
NM_029056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008939.mm.1 |
TC1200001252.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008943.mm.1 |
NM_029056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008943.mm.1 |
NM_029056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008943.mm.1 |
TC1200001252.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008957.mm.1 |
NM_029056.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008957.mm.1 |
NM_029056.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008957.mm.1 |
TC1200001252.mm.1 |
MTA TC |