Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Svip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160345.1 | NP_001153817.1 | 2 - 3 | 333 | 110 | |
NR_027704.1 | 2 - 3 | 224 | 110 | |||
GenBank mRNA | AK005663.1 | - | 2 - 3 | 202 | 110 | |
AK018351.1 | - | 2 - 3 | 164 | 110 | ||
AK029831.1 | - | 2 - 3 | 453 | 110 | ||
AK133737.1 | - | 2 - 3 | 142 | 110 | ||
AK158571.1 | - | 2 - 3 | 333 | 110 | ||
BC049720.1 | - | 208 | 110 | |||
BC100498.1 | - | 207 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |