Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200008461.mm.1 |
NM_001081457.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008461.mm.1 |
NM_012023.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008461.mm.1 |
TC1200001207.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008464.mm.1 |
NM_001081457.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008464.mm.1 |
NM_012023.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008464.mm.1 |
TC1200001207.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008465.mm.1 |
NM_001081457.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008465.mm.1 |
NM_012023.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008465.mm.1 |
TC1200001207.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008463.mm.1 |
NM_001081457.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008463.mm.1 |
NM_012023.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008463.mm.1 |
TC1200001207.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200008462.mm.1 |
NM_001081457.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008462.mm.1 |
NM_012023.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200008462.mm.1 |
TC1200001207.mm.1 |
MTA TC |