Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181820.2 | NP_861541.2 | 2 - 3 | 338 | 67 | |
GenBank mRNA | AK050105.1 | - | 1 - 2 | 233 | 67 | |
AY236493.1 | - | 2 - 3 | 301 | 67 | ||
BC040466.1 | - | 2 - 3 | 250 | 67 | ||
BC055939.1 | - | 1 - 2 | 67 | 67 | ||
BC106105.1 | - | 2 - 3 | 309 | 67 | ||
CB600173.1 | - | 2 - 3 | 350 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |