Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181820.2 | NP_861541.2 | 3 - 4 | 477 | 53 | |
GenBank mRNA | AK050105.1 | - | 2 - 3 | 372 | 53 | |
AY236493.1 | - | 3 - 4 | 440 | 53 | ||
AY263162.1 | - | 3 - 4 | 448 | 53 | ||
BC040466.1 | - | 3 - 4 | 389 | 53 | ||
BC055939.1 | - | 2 - 3 | 206 | 53 | ||
BC106105.1 | - | 3 - 4 | 448 | 53 | ||
CB600173.1 | - | 3 - 4 | 489 | 53 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |