Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Leng8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172736.3 | NP_766324.1 | 9 - 10 | 1464 | 87 | |
GenBank mRNA | AK034310.1 | - | 9 - 10 | 1469 | 87 | |
AK170629.1 | - | 9 - 10 | 1472 | 87 | ||
BC034448.1 | - | 1 - 2 | 188 | 87 | ||
BC041775.1 | - | 9 - 10 | 1596 | 87 | ||
BC042658.1 | - | 9 - 10 | 1452 | 5365041 | 87 | |
BC066768.1 | - | 9 - 10 | 1455 | 6407146 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |