Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Patl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026251.2 | NP_080527.2 | 9 - 10 | 1143 | 69 | |
GenBank mRNA | AK010811.1 | - | 2 - 3 | 215 | 69 | |
BC145646.1 | - | 7 - 8 | 924 | 40130949 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |