Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081006.1 | NP_001074475.1 | 14 - 15 | 3158 | 108 | |
NM_001177630.2 | NP_001171101.2 | 13 - 14 | 3256 | 108 | ||
NM_001177631.1 | NP_001171102.1 | 12 - 13 | 3151 | 108 | ||
NM_178059.5 | NP_835160.2 | 13 - 14 | 3364 | 108 | ||
GenBank mRNA | AB125594.2 | - | 13 - 14 | 3364 | 108 | |
AK147510.1 | - | 12 - 13 | 3058 | 109 | ||
BC026657.1 | - | 1 - 2 | 293 | 108 | ||
BC158051.1 | - | 13 - 14 | 2978 | 9007405 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |