Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081364.3 | NP_001074833.3 | 20 - 21 | 4297 | 77 | |
NM_001128084.2 | NP_001121556.2 | 21 - 22 | 4327 | 77 | ||
GenBank mRNA | AK122508.1 | - | 1938 | 77 | ||
BC076629.1 | - | 20 - 21 | 4297 | 30639338 | 77 | |
BC150741.1 | - | 21 - 22 | 4256 | 9087652 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |