Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sel1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039089.1 | NP_001034178.1 | 1 - 2 | 204 | 113 | |
NM_011344.2 | NP_035474.1 | 1 - 2 | 204 | 113 | ||
GenBank mRNA | AK005023.2 | - | 1 - 2 | 177 | 113 | |
AK005544.1 | - | 1 - 2 | 187 | 113 | ||
AK146022.1 | - | 1 - 2 | 178 | 113 | ||
AK160420.1 | - | 1 - 2 | 186 | 113 | ||
AK160936.1 | - | 1 - 2 | 162 | 113 | ||
AK161546.1 | - | 1 - 2 | 178 | 113 | ||
AK220502.1 | - | 1 - 2 | 204 | 113 | ||
BC026816.1 | - | 1 - 2 | 176 | 4457637 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |