Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpvl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289713.1 | NP_001276642.1 | 5 - 6 | 521 | 70 | |
NM_001289714.1 | NP_001276643.1 | 5 - 6 | 521 | 70 | ||
NM_027749.2 | NP_082025.1 | 5 - 6 | 607 | 70 | ||
GenBank mRNA | AK017087.1 | - | 5 - 6 | 522 | 70 | |
BC137839.1 | - | 5 - 6 | 596 | 8860858 | 70 | |
BC144966.1 | - | 5 - 6 | 510 | 9053514 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |