Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200027165.mm.1 |
NM_001113573.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027165.mm.1 |
NM_001113573.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027165.mm.1 |
NM_001113574.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027165.mm.1 |
NM_001113574.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027165.mm.1 |
NM_023336.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027165.mm.1 |
NM_023336.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027165.mm.1 |
TC0200003217.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200027175.mm.1 |
NM_001113573.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027175.mm.1 |
NM_001113573.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027175.mm.1 |
NM_001113574.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027175.mm.1 |
NM_001113574.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027175.mm.1 |
TC0200003217.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200027163.mm.1 |
NM_001113573.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027163.mm.1 |
NM_001113573.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027163.mm.1 |
NM_001113574.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027163.mm.1 |
NM_001113574.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027163.mm.1 |
NM_023336.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027163.mm.1 |
NM_023336.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027163.mm.1 |
TC0200003217.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200027174.mm.1 |
NM_001113573.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027174.mm.1 |
NM_001113573.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027174.mm.1 |
NM_001113574.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027174.mm.1 |
NM_001113574.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027174.mm.1 |
TC0200003217.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200027166.mm.1 |
NM_001113573.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027166.mm.1 |
NM_001113573.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027166.mm.1 |
NM_001113574.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027166.mm.1 |
NM_001113574.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027166.mm.1 |
NM_023336.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027166.mm.1 |
NM_023336.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027166.mm.1 |
TC0200003217.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200027167.mm.1 |
NM_001113573.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027167.mm.1 |
NM_001113573.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027167.mm.1 |
NM_001113574.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027167.mm.1 |
NM_001113574.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027167.mm.1 |
NM_023336.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027167.mm.1 |
NM_023336.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200027167.mm.1 |
TC0200003217.mm.1 |
MTA TC |