Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010695.1 | NP_034825.1 | 5 - 6 | 518 | 122 | |
GenBank mRNA | BC114544.1 | - | 5 - 6 | 517 | 40050061 | 122 |
D38581.1 | - | 5 - 6 | 518 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |