Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138942.3 | NP_620392.2 | 8 - 9 | 1391 | 86 | |
GenBank mRNA | AK132245.1 | - | 8 - 9 | 1392 | 86 | |
BC141022.1 | - | 8 - 9 | 1418 | 9055634 | 86 | |
BC171949.1 | - | 8 - 9 | 1418 | 9054235 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |