Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rexo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207234.2 | NP_997117.2 | 1 - 2 | 331 | 135 | |
GenBank mRNA | AB221675.1 | - | 1 - 2 | 263 | 135 | |
AK133982.1 | - | 1 - 2 | 253 | 136 | ||
AK138894.1 | - | 1 - 2 | 246 | 135 | ||
BC060147.1 | - | 1 - 2 | 257 | 6414260 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |