Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fnbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038700.2 | NP_001033789.1 | 11 - 12 | 1436 | 57 | |
NM_001177648.1 | NP_001171119.1 | 13 - 14 | 1634 | 57 | ||
NM_001177649.1 | NP_001171120.1 | 11 - 12 | 1436 | 57 | ||
NM_001177650.1 | NP_001171121.1 | 10 - 11 | 1286 | 57 | ||
GenBank mRNA | AK122308.1 | - | 13 - 14 | 1634 | 57 | |
AK143512.1 | - | 10 - 11 | 1287 | 57 | ||
AK157135.1 | - | 11 - 12 | 1427 | 57 | ||
AK170497.1 | - | 11 - 12 | 1386 | 57 | ||
AK172100.1 | - | 11 - 12 | 1380 | 57 | ||
U40751.1 | - | 3 - 4 | 281 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |