Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cts7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019539.3 | NP_062412.1 | 1 - 2 | 206 | 75 | |
GenBank mRNA | AK017466.1 | - | 1 - 2 | 232 | 75 | |
BC050198.1 | - | 1 - 2 | 219 | 75 | ||
BC064740.1 | - | 1 - 2 | 206 | 30018572 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |