Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008498.2 | NP_001008498.2 | 4 - 5 | 717 | 65 | |
GenBank mRNA | AK014304.1 | - | 4 - 5 | 716 | 65 | |
BC085113.1 | - | 4 - 5 | 673 | 30730999 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |