Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175238.5 | NP_780447.4 | 26 - 27 | 3127 | 142 | |
GenBank mRNA | AY428571.2 | - | 26 - 27 | 3111 | 142 | |
AY585206.1 | - | 25 - 26 | 3120 | 142 | ||
JX255737.1 | - | 25 - 26 | 3063 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |