Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175238.5 | NP_780447.4 | 8 - 9 | 854 | 86 | |
GenBank mRNA | AY428571.2 | - | 8 - 9 | 838 | 86 | |
AY585206.1 | - | 7 - 8 | 847 | 86 | ||
JX255737.1 | - | 7 - 8 | 790 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |