Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rc3h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100591.1 | NP_001094061.1 | 8 - 9 | 1333 | 80 | |
NM_001290642.1 | NP_001277571.1 | 8 - 9 | 1333 | 80 | ||
GenBank mRNA | AK053071.1 | - | 8 - 9 | 1333 | 80 | |
BC117951.1 | - | 8 - 9 | 1326 | 40098089 | 80 | |
BC117952.1 | - | 8 - 9 | 1326 | 40098090 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |