Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033960.1 | NP_001029132.1 | 8 - 9 | 1186 | 97 | |
NM_146121.2 | NP_666233.2 | 8 - 9 | 1310 | 97 | ||
GenBank mRNA | AK021286.1 | - | 8 - 9 | 1225 | 97 | |
AK029667.1 | - | 8 - 9 | 1172 | 97 | ||
AK044346.1 | - | 8 - 9 | 1187 | 97 | ||
AK220489.1 | - | 7 - 8 | 1131 | 97 | ||
BC138337.1 | - | 8 - 9 | 1212 | 8861357 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |