Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tanc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290659.1 | NP_001277588.1 | 12 - 13 | 1992 | 76 | |
NM_198294.3 | NP_938036.2 | 13 - 14 | 2168 | 76 | ||
GenBank mRNA | AB093299.1 | - | 2 - 3 | 261 | 76 | |
AK030022.1 | - | 10 - 11 | 1772 | 76 | ||
BC079914.1 | - | 12 - 13 | 1971 | 30668628 | 76 | |
BC141367.1 | - | 12 - 13 | 1982 | 9055980 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |