Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pacs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081170.1 | NP_001074639.1 | 14 - 15 | 1565 | 98 | |
NM_001291444.1 | NP_001278373.1 | 14 - 15 | 1571 | 98 | ||
NM_001291445.1 | NP_001278374.1 | 13 - 14 | 1475 | 98 | ||
GenBank mRNA | AK030003.1 | - | 9 - 10 | 934 | 98 | |
AK036581.1 | - | 13 - 14 | 1451 | 98 | ||
AK054000.1 | - | 14 - 15 | 1546 | 98 | ||
AK122326.1 | - | 12 - 13 | 1254 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |