Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pacs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081170.1 | NP_001074639.1 | 8 - 9 | 859 | 74 | |
NM_001291444.1 | NP_001278373.1 | 8 - 9 | 859 | 74 | ||
NM_001291445.1 | NP_001278374.1 | 7 - 8 | 769 | 74 | ||
GenBank mRNA | AK030003.1 | - | 3 - 4 | 227 | 74 | |
AK054000.1 | - | 8 - 9 | 834 | 74 | ||
AK122326.1 | - | 6 - 7 | 548 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |