Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081033.1 | NP_001074502.1 | 10 - 11 | 1837 | 79 | |
GenBank mRNA | AK042383.1 | - | 7 - 8 | 615 | 79 | |
BC158125.1 | - | 10 - 11 | 1977 | 9088197 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |