Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trub2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290496.1 | NP_001277425.1 | 4 - 5 | 436 | 58 | |
NM_145520.5 | NP_663495.3 | 4 - 5 | 436 | 58 | ||
GenBank mRNA | AK043949.1 | - | 4 - 5 | 431 | 58 | |
AK087102.1 | - | 4 - 5 | 418 | 58 | ||
AK090004.1 | - | 1268 | 58 | |||
AK167294.1 | - | 4 - 5 | 412 | 58 | ||
BC015285.1 | - | 4 - 5 | 415 | 4239390 | 58 | |
BC024075.1 | - | 4 - 5 | 405 | 5097466 | 58 | |
BU053791.1 | - | 4 - 5 | 388 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |