Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gimap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008376.3 | NP_032402.2 | 1 - 2 | 112 | 74 | |
GenBank mRNA | AK156732.1 | - | 1 - 2 | 107 | 74 | |
BC132601.1 | - | 1 - 2 | 72 | 40130878 | 74 | |
BC132603.1 | - | 1 - 2 | 72 | 40130880 | 74 | |
BY095291.1 | - | 1 - 2 | 113 | 74 | ||
BY751570.1 | - | 1 - 2 | 107 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |