Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012518.3 | NP_001012536.2 | 25 - 26 | 3567 | 77 | |
NM_001109686.2 | NP_001103156.1 | 24 - 25 | 3408 | 77 | ||
NM_001109687.2 | NP_001103157.1 | 24 - 25 | 3423 | 77 | ||
NM_172545.4 | NP_766133.2 | 25 - 26 | 3546 | 77 | ||
GenBank mRNA | AB205007.1 | - | 25 - 26 | 3573 | 77 | |
AK049454.1 | - | 22 - 23 | 2817 | 77 | ||
AK052174.1 | - | 25 - 26 | 3530 | 77 | ||
AK082062.1 | - | 25 - 26 | 3557 | 77 | ||
AK129461.1 | - | 6 - 7 | 658 | 77 | ||
AK149125.1 | - | 5 - 6 | 490 | 77 | ||
BC023904.1 | - | 7 - 8 | 812 | 77 | ||
BC056938.1 | - | 22 - 23 | 3124 | 77 | ||
BC089302.1 | - | 22 - 23 | 3276 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |