Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083810.2 | NP_001077279.1 | 7 - 8 | 983 | 90 | |
NM_001110849.1 | NP_001104319.1 | 8 - 9 | 846 | 90 | ||
NM_175181.5 | NP_780390.3 | 7 - 8 | 932 | 90 | ||
GenBank mRNA | AK011178.1 | - | 2 - 3 | 180 | 90 | |
AK030499.1 | - | 7 - 8 | 891 | 89 | ||
AK145981.1 | - | 7 - 8 | 934 | 90 | ||
BC048394.1 | - | 7 - 8 | 842 | 6391924 | 90 | |
BC172132.1 | - | 6 - 7 | 658 | 9054802 | 90 | |
BC172133.1 | - | 6 - 7 | 658 | 90 | ||
BC172158.1 | - | 5 - 6 | 566 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |