Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snapc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290419.1 | NP_001277348.1 | 19 - 20 | 2545 | 60 | |
NM_172339.4 | NP_758842.1 | 18 - 19 | 2426 | 60 | ||
GenBank mRNA | AK077522.1 | - | 18 - 19 | 2393 | 60 | |
BC044754.2 | - | 3 - 4 | 819 | 60 | ||
BC057031.1 | - | 19 - 20 | 2511 | 6833968 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |