Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cercam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207298.2 | NP_997181.1 | 10 - 11 | 1361 | 76 | |
GenBank mRNA | AK155384.1 | - | 10 - 11 | 1357 | 76 | |
BC052176.1 | - | 10 - 11 | 1324 | 76 | ||
BC138847.1 | - | 10 - 11 | 1340 | 8861869 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |