Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164314.1 | NP_001157786.1 | 3 - 4 | 494 | 85 | |
NM_001164488.1 | NP_001157960.1 | 3 - 4 | 427 | 85 | ||
NM_011710.3 | NP_035840.3 | 3 - 4 | 440 | 85 | ||
GenBank mRNA | AK004541.1 | - | 3 - 4 | 428 | 85 | |
AK007754.1 | - | 1 - 2 | 56 | 85 | ||
AK150090.1 | - | 3 - 4 | 458 | 85 | ||
AK150267.1 | - | 4 - 5 | 542 | 85 | ||
AK152966.1 | - | 3 - 4 | 458 | 85 | ||
AK153337.1 | - | 3 - 4 | 466 | 84 | ||
BC003450.1 | - | 3 - 4 | 426 | 2654231 | 85 | |
BC046232.1 | - | 3 - 4 | 433 | 6813406 | 85 | |
CD563585.1 | - | 3 - 4 | 414 | 85 | ||
CT010258.1 | - | 2 - 3 | 326 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |