Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Celf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244891.1 | NP_001231820.1 | 8 - 9 | 993 | 62 | |
NM_001244903.1 | NP_001231832.1 | 7 - 8 | 940 | 62 | ||
NM_017368.3 | NP_059064.2 | 8 - 9 | 1092 | 62 | ||
NM_198683.2 | NP_941955.1 | 7 - 8 | 940 | 62 | ||
GenBank mRNA | AB050499.1 | - | 4 - 5 | 401 | 62 | |
AF267535.1 | - | 6 - 7 | 696 | 62 | ||
AF314172.1 | - | 6 - 7 | 534 | 62 | ||
AJ007987.1 | - | 6 - 7 | 685 | 62 | ||
AK014492.1 | - | 7 - 8 | 853 | 62 | ||
AK143698.1 | - | 8 - 9 | 894 | 62 | ||
AK156725.1 | - | 7 - 8 | 853 | 62 | ||
AK161213.1 | - | 6 - 7 | 1710 | 62 | ||
AK218377.1 | - | 3 - 4 | 265 | 62 | ||
BC021393.1 | - | 7 - 8 | 805 | 62 | ||
X61451.1 | - | 3 - 4 | 388 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |