Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgkz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166597.1 | NP_001160069.1 | 13 - 14 | 1793 | 115 | |
NM_138306.2 | NP_612179.2 | 12 - 13 | 1192 | 115 | ||
GenBank mRNA | AK087503.1 | - | 13 - 14 | 1794 | 115 | |
AK170936.1 | - | 9 - 10 | 815 | 115 | ||
BC049228.1 | - | 12 - 13 | 1228 | 3668704 | 115 | |
BQ443200.1 | - | 3 - 4 | 196 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |