Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exosc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144886.2 | NP_659135.1 | 5 - 6 | 432 | 71 | |
GenBank mRNA | AK144234.1 | - | 5 - 6 | 441 | 71 | |
AK166881.1 | - | 5 - 6 | 442 | 71 | ||
BC021485.1 | - | 5 - 6 | 456 | 5344620 | 71 | |
BC021807.1 | - | 5 - 6 | 454 | 3980155 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |