Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sel1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033296.2 | NP_001028468.1 | 18 - 19 | 1891 | 87 | |
GenBank mRNA | AK016809.1 | - | 18 - 19 | 1837 | 87 | |
AK132650.1 | - | 18 - 19 | 1892 | 87 | ||
BC150948.1 | - | 18 - 19 | 1863 | 9087859 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |