Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sel1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033296.2 | NP_001028468.1 | 1 - 2 | 142 | 125 | |
GenBank mRNA | AK016809.1 | - | 1 - 2 | 159 | 125 | |
AK131753.1 | - | 1 - 2 | 141 | 125 | ||
AK132650.1 | - | 1 - 2 | 143 | 125 | ||
BC150948.1 | - | 1 - 2 | 114 | 9087859 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |