Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp8b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080944.3 | NP_001074413.2 | 23 - 24 | 2809 | 96 | |
GenBank mRNA | AK155448.1 | - | 23 - 24 | 2819 | 96 | |
AK169967.1 | - | 23 - 24 | 2837 | 96 | ||
BC130260.1 | - | 5 - 6 | 783 | 96 | ||
BC152337.1 | - | 5 - 6 | 783 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |