Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp8b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080944.3 | NP_001074413.2 | 7 - 8 | 544 | 130 | |
GenBank mRNA | AK040903.1 | - | 8 - 9 | 713 | 130 | |
AK086607.1 | - | 7 - 8 | 578 | 130 | ||
AK155448.1 | - | 7 - 8 | 555 | 130 | ||
AK169967.1 | - | 7 - 8 | 567 | 130 | ||
AK173307.1 | - | 7 - 8 | 544 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Mus musculus Probe spans exons |