Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024616.1 | NP_001019787.2 | 11 - 12 | 1729 | 131 | |
GenBank mRNA | AK045520.1 | - | 11 - 12 | 1751 | 131 | |
AK081440.1 | - | 11 - 12 | 1739 | 131 | ||
AK157020.1 | - | 11 - 12 | 1764 | 131 | ||
AK162461.1 | - | 7 - 8 | 1109 | 131 | ||
AK171643.1 | - | 11 - 12 | 1740 | 131 | ||
BC006855.1 | - | 3 - 4 | 3461 | 131 | ||
BC020111.1 | - | 11 - 12 | 1737 | 131 | ||
BC021476.1 | - | 11 - 12 | 1744 | 131 | ||
BC023692.1 | - | 11 - 12 | 1757 | 131 | ||
BC040791.1 | - | 3 - 4 | 3461 | 131 | ||
BC051138.1 | - | 11 - 12 | 1743 | 131 | ||
BC058585.1 | - | 11 - 12 | 1742 | 131 | ||
BC085507.1 | - | 11 - 12 | 1729 | 5716088 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |