Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brsk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003920.3 | NP_001003920.2 | 8 - 9 | 974 | 81 | |
NM_001168572.1 | NP_001162044.1 | 8 - 9 | 974 | 81 | ||
GenBank mRNA | AY533671.1 | - | 8 - 9 | 816 | 81 | |
BC086636.1 | - | 8 - 9 | 973 | 6402113 | 81 | |
EU016556.1 | - | 8 - 9 | 822 | 81 | ||
EU016557.1 | - | 8 - 9 | 816 | 81 | ||
EU016558.1 | - | 8 - 9 | 816 | 81 | ||
EU586326.1 | - | 8 - 9 | 1004 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |