Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Duox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099297.1 | NP_001092767.1 | 9 - 10 | 1149 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |