Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gas7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109657.3 | NP_001103127.1 | 12 - 13 | 1347 | 89 | |
NM_001277079.1 | NP_001264008.1 | 12 - 13 | 1176 | 89 | ||
NM_001277080.1 | NP_001264009.1 | 11 - 12 | 1090 | 89 | ||
NM_008088.5 | NP_032114.2 | 12 - 13 | 1153 | 89 | ||
GenBank mRNA | AF133184.1 | - | 8 - 9 | 1025 | 89 | |
AK017394.1 | - | 12 - 13 | 1146 | 89 | ||
AK076926.1 | - | 13 - 14 | 1229 | 89 | ||
AK152149.1 | - | 10 - 11 | 850 | 89 | ||
AK154463.1 | - | 12 - 13 | 1150 | 89 | ||
AK172943.1 | - | 8 - 9 | 969 | 89 | ||
BC120528.1 | - | 12 - 13 | 1074 | 40129451 | 89 | |
BC120530.1 | - | 12 - 13 | 1074 | 40129453 | 89 | |
CV562268.1 | - | 4 - 5 | 391 | 89 | ||
U19860.1 | - | 12 - 13 | 1179 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |