Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap4e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175550.3 | NP_780759.2 | 17 - 18 | 2389 | 75 | |
GenBank mRNA | AK036944.1 | - | 17 - 18 | 2370 | 75 | |
AK144636.1 | - | 17 - 18 | 2370 | 75 | ||
BC042530.1 | - | 4 - 5 | 580 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |