Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap4e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175550.3 | NP_780759.2 | 6 - 7 | 743 | 101 | |
GenBank mRNA | AK036944.1 | - | 6 - 7 | 724 | 101 | |
AK039257.1 | - | 6 - 7 | 731 | 101 | ||
AK143351.1 | - | 6 - 7 | 733 | 101 | ||
AK144636.1 | - | 6 - 7 | 723 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |